LIF009183339 Life Chemicals Inc.

4,8-Dimethyl-2-oxo-3,4-dihydro-2H-pyrido[1,2-a]pyrimidine-4-carboxylic acid

  • MDL number MFCD01304083
  • Cas Number : 393802-66-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart