LIF009184141 Life Chemicals Inc.

3-Methyl-2-(thiophene-2-sulfonylamino)-butyric acid

  • MDL number MFCD03834403
  • Cas Number : 82068-14-2
  • Molecular Weight : 263.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart