LIF009184204 Life Chemicals Inc.

3-(3,5-Dimethyl-1H-pyrazol-4-yl)propanoic acid

  • MDL number MFCD03030172
  • Cas Number : 890625-93-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart