LIF009187302 Life Chemicals Inc.

7-Bromo-1,5-dimethyl-1H-indole-2,3-dione

  • MDL number MFCD00566037
  • Cas Number : 124807-89-2
  • Molecular Weight : 254.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart