LIF009209189 Life Chemicals Inc.

6-(Morpholin-4-ylsulfonyl)-1,3-benzothiazol-2-amine

  • MDL number MFCD03450653
  • Cas Number : 440627-31-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart