LIF009209906 Life Chemicals Inc.

1-Benzyl-1H-benzimidazole-2-carbaldehyde

  • MDL number MFCD03130245
  • Cas Number : 180000-91-3
  • Molecular Weight : 236.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart