LIF009209987 Life Chemicals Inc.

6-Methoxy-quinolin-8-ylamine

  • MDL number MFCD00672902
  • Cas Number : 90-52-8
  • Molecular Weight : 174.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart