LIF009210104 Life Chemicals Inc.

(4-Methyl-benzothiazol-2-yl)-hydrazine

  • MDL number MFCD04448793
  • Cas Number : 20174-68-9
  • Molecular Weight : 179.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart