LIF009210108 Life Chemicals Inc.

(4-Methoxy-benzothiazol-2-yl)-hydrazine

  • MDL number MFCD04448795
  • Cas Number : 53065-23-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart