LIF009210111 Life Chemicals Inc.

(4-Chloro-benzothiazol-2-yl)-hydrazine

  • MDL number MFCD04448798
  • Cas Number : 51769-38-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart