LIF009210114 Life Chemicals Inc.

6-Ethyl-2-hydrazino-1,3-benzothiazole

  • MDL number MFCD04971774
  • Cas Number : 890091-90-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart