LIF009210117 Life Chemicals Inc.

(6-Methoxy-benzothiazol-2-yl)-hydrazine

  • MDL number MFCD01439956
  • Cas Number : 20174-70-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart