LIF009210120 Life Chemicals Inc.

(6-Chloro-benzothiazol-2-yl)-hydrazine

  • MDL number MFCD00832729
  • Cas Number : 51011-54-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart