LIF009210129 Life Chemicals Inc.

(4,6-Dimethyl-benzothiazol-2-yl)-hydrazine

  • MDL number MFCD04448811
  • Cas Number : 80945-72-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart