LIF009210134 Life Chemicals Inc.

2-Hydrazino-4,7-dimethoxy-1,3-benzothiazole

  • MDL number MFCD04448815
  • Cas Number : 872696-10-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart