LIF009210173 Life Chemicals Inc.

2-Chloro-4-methyl-benzothiazole

  • MDL number MFCD03840701
  • Cas Number : 3622-32-0
  • Molecular Weight : 183.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart