LIF009210177 Life Chemicals Inc.

2-Chloro-4-methoxy-benzothiazole

  • MDL number MFCD04448819
  • Cas Number : 3507-27-5
  • Molecular Weight : 199.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart