LIF009210180 Life Chemicals Inc.

2,4-Dichloro-benzothiazole

  • MDL number MFCD00127716
  • Cas Number : 3622-30-8
  • Molecular Weight : 204.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart