LIF009210188 Life Chemicals Inc.

2-Chloro-6-fluoro-benzothiazole

  • MDL number MFCD04448822
  • Cas Number : 399-74-6
  • Molecular Weight : 187.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart