LIF009210192 Life Chemicals Inc.

2,4,6-Trichloro-benzothiazole

  • MDL number MFCD00043998
  • Cas Number : 25745-12-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart