LIF009210194 Life Chemicals Inc.

2,5-Dichloro-4-methyl-1,3-benzothiazole

  • MDL number MFCD04971825
  • Cas Number : 887204-62-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart