LIF009210196 Life Chemicals Inc.

2,7-Dichloro-4-methyl-1,3-benzothiazole

  • MDL number MFCD04971827
  • Cas Number : 80945-85-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart