LIF009210197 Life Chemicals Inc.

2-Chloro-4,5-dimethyl-1,3-benzothiazole

  • MDL number MFCD04971828
  • Cas Number : 252681-54-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart