LIF009210199 Life Chemicals Inc.

2-Chloro-4,7-dimethyl-1,3-benzothiazole

  • MDL number MFCD04971830
  • Cas Number : 80945-84-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart