LIF009210201 Life Chemicals Inc.

2-Chloro-4-methoxy-7-methyl-benzothiazole

  • MDL number MFCD04971832
  • Cas Number : 108773-00-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart