LIF009210204 Life Chemicals Inc.

2-Chloro-4,6-difluoro-1,3-benzothiazole

  • MDL number MFCD04971835
  • Cas Number : 252681-57-5
  • Molecular Weight : 205.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart