LIF009210205 Life Chemicals Inc.

6-Chloro[1,3]dioxolo[4,5-f][1,3]benzothiazole

  • MDL number MFCD04448823
  • Cas Number : 155559-77-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart