LIF009210207 Life Chemicals Inc.

4-ethyl-1,3-benzothiazol-2-amine

  • MDL number MFCD04971837
  • Cas Number : 139331-68-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart