LIF009210208 Life Chemicals Inc.

4-isopropylbenzo[d]thiazol-2-amine

  • MDL number MFCD04971838
  • Cas Number : 1024490-13-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart