LIF009210218 Life Chemicals Inc.

5-chloro-4-methyl-1,3-benzothiazol-2-amine

  • MDL number MFCD04971842
  • Cas Number : 65373-18-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart