LIF009210224 Life Chemicals Inc.

4-Methoxy-7-methyl-benzothiazol-2-ylamine

  • MDL number MFCD04448826
  • Cas Number : 88686-30-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart