LIF009210244 Life Chemicals Inc.

3-Imidazo[1,2-a]pyridin-2-yl-phenylamine

  • MDL number MFCD00702245
  • Cas Number : 313231-71-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart