LIF009210269 Life Chemicals Inc.

1-Benzyl-pyrimidine-2,4,6-trione

  • MDL number MFCD00104232
  • Cas Number : 91360-95-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart