LIF009210271 Life Chemicals Inc.

[3-(8-Methylimidazo[1,2-a]pyridin-2-yl)phenyl]amine

  • MDL number MFCD04971850
  • Cas Number : 878437-60-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart