LIF019469401 Life Chemicals Inc.

4-(Pyrrolidine-1-sulfonyl)-phenylamine

  • MDL number MFCD00277212
  • Cas Number : 88327-91-7
  • Molecular Weight : 226.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart