LIF019472671 Life Chemicals Inc.

4-(4-Methyl-piperazine-1-sulfonyl)-phenylamine

  • MDL number MFCD03050283
  • Cas Number : 21623-68-7
  • Molecular Weight : 255.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart