LIF019472792 Life Chemicals Inc.

2-(methoxymethyl)-1H-1,3-benzodiazole

  • MDL number MFCD01056151
  • Cas Number : 7146-97-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart