LIF019474420 Life Chemicals Inc.

1-Amino-3-(4-chloro-3-methyl-phenoxy)-propan-2-ol

  • MDL number MFCD04444357
  • Cas Number : 71954-32-0
  • Molecular Weight : 215.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart