LIF019492424 Life Chemicals Inc.

(2,6-Dioxo-1,2,3,6-tetrahydro-pyrimidin-4-yl)-acetic acid ethyl ester

  • MDL number MFCD01106346
  • Cas Number : 6426-84-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart