LIF019492481 Life Chemicals Inc.

4-(4-Fluorophenyl)-4,5,6,7-tetrahydro-3H-imidazo[4,5-c]pyridine

  • MDL number MFCD00841387

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart