LIF019492482 Life Chemicals Inc.

4-Phenyl-4,5,6,7-tetrahydro-3H-imidazo[4,5-c]pyridine-6-carboxylic acid

  • MDL number MFCD00488388
  • Cas Number : 178456-18-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart