LIF019492486 Life Chemicals Inc.

5-(Anilinocarbonothioyl)-4,5,6,7-tetrahydro-3H-imidazo[4,5-c]pyridine- 6-carboxylic acid hydrochloride

  • MDL number MFCD04448843
  • Cas Number : 1030285-50-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart