LIF019493495 Life Chemicals Inc.

1-Phenyladamantane

  • Cas Number : 780-68-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart