LIF019493559 Life Chemicals Inc.

(2,4-Dioxo-3-propyl-1,3-thiazolidin-5-yl)acetic acid

  • MDL number MFCD04971539
  • Cas Number : 685861-79-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart