LIF019493579 Life Chemicals Inc.

2-[(4-hydroxyphenyl)formamido]acetic acid

  • MDL number MFCD00237117
  • Cas Number : 2482-25-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart