LIF019493610 Life Chemicals Inc.

4-Amino-2-oxo-1,2-dihydro-pyrimidine-5-carbonitrile

  • MDL number MFCD00767571
  • Cas Number : 16462-28-5
  • Molecular Weight : 136.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart