LIF019493617 Life Chemicals Inc.

9H-Carbazol-3-ylamine

  • MDL number MFCD00022213

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart