LIF019493645 Life Chemicals Inc.

(4-Methoxypyridin-3-yl)boronic acid hydrochloride

  • MDL number MFCD04113800
  • Cas Number : 874959-97-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart