LIF019493682 Life Chemicals Inc.

Spiro[Chromene-2,4'-piperidin]-4(3H)-one hydrochloride

  • MDL number MFCD04971902
  • Cas Number : 159635-39-9
  • Molecular Weight : 253.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart