LIF019493727 Life Chemicals Inc.

(3-Phenyl-1-adamantyl)acetic acid

  • Cas Number : 161036-56-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart